Learn More
Cayman Chemical 22 S 23 S-HomobrasInolIde 5mg
SDP
Supplier:  Cayman Chemical 328845 MG
Encompass_Preferred
A synthetic brassinosteroid; increases the rooting percentage in P. abies cuttings when applied topically at 60 ppm
Catalog No. 50-295-0486