Learn More
Electron Microscopy Sciences 22 Spin Bars Assortment, Round
SDP
Supplier:  Electron Microscopy Sciences 6451022
Encompass_Preferred
22 round magnetic stirring bars. PTFE with removable pivot ring . Includes 2 each: 1/2"x3/4", 1"x 5/16", 2"x 3/8", 1/2"x 5/16", 3/4"x 5/16", 1"x 3/8", 1 1/2"x 5/16", 1 3/4"x 5/16", 2"x 5/16" and 1 each: 1/4"x 5/16", 1/4"x 3/8", 11/2"x3/8", 1 3/4"x.
Catalog No. 50-270-16