Learn More
TARGETMOL CHEMICALS INC 22393-62-0-peak 2 1MG
SDP
Supplier:  TARGETMOL CHEMICALS INC T97961MG
Encompass_Preferred
Also available in 5 mg and bulk. Please contact Fisher for quotes. 22393-62-0-peak 2 exhibits some aspects of the antiestrogenic activity and other actions that may be connected to the estrogenic properties. A mixture of the (Z)- and (E)-isomers (Broparestrol, INN) is used in dermatology. Purity 98.71%
Catalog No. 50-288-0192