Learn More
Access Technologies Huber Point Non-Coring Needles, 22ga, 3/4” (1.6cm), Black, Needleless Injector Sites
Supplier:  Access Technologies PG2275BC
Encompass
Feature an architecturally contoured “B” bevel needle tip and a unique knarled hub. This hub provides positive grip action when attaching the needle to a syringe or luer lock. The PosiGrip hub is color coded for easy gauge identification.
Catalog No. NC9981841
 Restricted Item