Learn More
Thomas Scientific 22"D Combo Upright Freezer Drawer Racks for 15ml and 50ml Tubes, 22"L x 4 15/16"H x 5 1/2"W, Holds 80 (15ml) and 39 (50ml) Tubes.
SDP
Supplier:  Thomas Scientific 1148W79
Encompass
Racks for specially designed to provide an ideal alternative storing large quantity of samples in 15ml and 50ml centrifuge tubes. Constructed of corrosion resistant stainsteel. Drawer Racks facilitate both access to and storage of samples without removing the whole racks from the freezer.
Catalog No. 50-196-048