Learn More
Ace Glass, Inc. 22mm x 41cm chromatography column
SDP
Supplier:  Ace Glass, Inc. 588839
Encompass_Preferred
22mm x 41cm chromatography column with 2mm bore 1:5 taper solid PTFE stopcock plug at the bottom.
Catalog No. 50-978-589