Learn More
Medchemexpress LLC 22R-Hydroxycholester 1mg | 17954-98-2 | 402.65 g·mol⁻¹ | C27H46O2 | 1 MG

Supplier: Medchemexpress LLC HY1169691MG
22(R)-Hydroxycholesterol is an oxysterol and an endogenous agonist of the liver X receptor (LXR). Supplied as a purified research-grade powder, it is used to study LXR-mediated pathways, cholesterol metabolism, and related conditions such as Tangier disease. Store the powder at -20°C; in solution keep at -80°C for long-term storage. Safety data and handling information are available in the product SDS.
- Endogenous LXR agonist useful for receptor and cholesterol metabolism studies.
- High reported purity (approximately 99.8%).
- Chemical formula C27H46O2 and molecular weight ~402.65 g·mol⁻¹.
- Supplied as a stable powder suitable for long-term storage at -20°C.
- Intended for research use only; consult SDS for safety and handling.
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.