Learn More
Dazor Mfg Corporation Circline 22W Fluorescent Bulb (Daylight) Mininum order 3 ea
Supplier:  Dazor Mfg Corporation FC8T9/DL
Encompass_Preferred
This item has a minimum qty of 2 per supplier requirements.
Replacement 22W Daylight fluorescent bulb for Circline Magnifiers Product DetailsBulb Type: FC8T9; Bulb Category: Fluorescent ; Bulb Style: Circline; Color Temperature: Daylight; CCT: 6500K; Brightness: 925 lumens; CRI: 75; Pin Base: 4-pin G10q; Tube Diameter: 1.125 inches; Bulb Diameter: 8.25 inches; Average Bulb Life: 12,000 hours
Catalog No. NC0653071