Learn More
Electron Microscopy Sciences Microscope Slide Cover Glass #1, 22x50 mm, 70 per oz 1 OZ
SDP
Supplier:  Electron Microscopy Sciences 7220041
Encompass_Preferred
Manufactured from clear white, borosilicate glass. They offer super clarity, are perfectly flat, and corrosion resistant. All cover glasses are uniform in size and thickness.
Size: 22 x 50 mm
Package: 70 per oz
Thickness: .13 - .17mm
Catalog No. NC1420323
This item has been discontinued by the Fisher Scientific channel. Please to view product availability in your area.