Learn More
Selleck Chemical LLC 23-cGAMP Sodium Salt 1mg
SDP
Supplier:  Selleck Chemical LLC S79041MG
Encompass_Preferred
2',3'-cGAMP Sodium Salt (2'-3'-cyclic GMP-AMP Sodium) is produced in response to DNA in the cytoplasm in mammalian cells and bindsSTING with high affinity and is an effective inducer ofinterferon-? (IFN?). 2',3'-cGAMP binds toSTING with Kd of 3.79 nM. *For Research & Development use only.  Product is not intended for drug, household, or human consumption.
Catalog No. 50-217-3322
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.