Learn More
Aviva Systems Biology Corp 2310028H24Rik Antibody - N-terminal region (ARP55477_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP55477_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against 2310028H24Rik. It was validated on Western Blot. . Alternate names: Fam219a, 2310028H24RikApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Rabbit, Rat, Zebrafish
Catalog No. 50-286-042