Learn More
Chemglass Life Sciences Chemglass Metal Luer to 24/40 Inner Joint Adapter - connects 1/8 in PTFE tubing to 24/40 tapered glass joint
SDP
Supplier:  Chemglass Life Sciences CG105504
Encompass_Preferred
The Chemglass CG-1055-04 metal luer to 24/40 inner joint adapter provides a secure connection between 1/8 in (3.18 mm) OD PTFE tubing and standard 24/40 tapered glass joints. It is designed for use with vacuum manifolds and other air-sensitive transfer applications where a reliable luer-to-glass seal is required.
  • Connects 1/8 in (3.18 mm) OD PTFE tubing to a 24/40 inner taper joint.
  • Metal luer for durable, leak-resistant connections.
  • Suitable for vacuum and air-sensitive transfer applications.
  • Sold individually (each).
Catalog No. 50-273-3600