Learn More
Uline 6 Mil Poly Bags, Clear, 100
Supplier:  Uline S11040
Encompass
Eliminate tears punctures and broken bags,Recommended for use with heavy metal parts,Virgin polyethylene film,Meets FDA and USDA specifications,LowLinear Low Density,Sulfur free,Amine and Silicone free,Latex free,BHT free Butylated Hydroxy Toluene,Chlorine free,PVC free,TEMPERATURE 20 - 150 F,Recyclable 4
Catalog No. NC1766804