Learn More
Medchemexpress LLC HY-N2370 2mg Medchemexpress, 24-Hydroxycholesterol CAS:30271-38-6 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HYN23702MG
Encompass_Preferred
Medchemexpress, HY-N2370 2mg 24-Hydroxycholesterol CAS:30271-38-6 24-Hydroxycholesterol is a natural sterol, which serves as a positive allosteric modulator of N-Methyl-d-Aspartate (NMDA) receptorsR , and a potent activator of the transcription factors LXR . Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-193-3922