Learn More
Medchemexpress LLC 24-Hydroxycholesterol | 30271-38-6 | 50 MG
SDP
Supplier:  Medchemexpress LLC HYN237050MG
Encompass_Preferred
24-Hydroxycholesterol is a natural sterol. It functions as a positive allosteric modulator of N-Methyl-d-Aspartate (NMDA) receptors and is a potent activator of the transcription factors LXR. This product is intended for research use only.
  • Positive allosteric modulator of N-Methyl-d-Aspartate (NMDA) receptors.
  • Potent activator of the transcription factors LXR.
  • Intended for research use only.
Catalog No. 50-004-21391