Learn More
US Plastic Corporation 24 oz. Wide Mouth Bottle with Red and White Sprayer
SDP
Supplier:  US Plastic Corporation 66631
Encompass
The evolution of trigger sprayers. Fill without spills, larger opening than a gallon jug. 45/400 neck finish. Output .85cc per stroke. Fully adjustable spray pattern. Sprayers packed completely assembled. Bottles have a black graduation scale silk-screened.
Catalog No. NC0374151
Item Discontinued This item has been discontinued by the supplier. Please to view product availability in your area.