Learn More
ALPAQUA ENGINEERING LLC 24 POSITION SBS TUBE RACK
Supplier:  ALPAQUA ENGINEERING LLC A000280
Encompass
24 position SBS-compatible tube rack. Can be customized for tube diameter and height
Catalog No. NC2294008