Learn More
Enzo Life Sciences 24(S)-Hydroxycholesterol ELISA kit (1x96wells)
SDP
Supplier:  Enzo Life Sciences ADI9002100001
Encompass_Preferred
The 24(S)-Hydroxycholesterol ELISA kit is a complete, colorimetric, competitive immunoassay kit for the quantitative determination of 24(S)-Hydroxycholesterol in tissue culture media (culture supernatant), cerebral spinal fluid and tissue homogenate samples with results in just 2 hours. Assay Time: 2 hours. Applications: ELISA, Colorimetric detection . Species reactivity: Species independent .
Catalog No. 50-200-6868