Learn More
Sterlitech Corporation cellQART 24-Well Cell Culture Insert PET, 1.0um, Translucent, 48/pk
SDP
Supplier:  Sterlitech Corporation 9321002
Encompass_Preferred
cellQART 24-Well Cell Culture Insert PET, 1.0um, Translucent, 48/pk
Catalog No. NC2004017