Learn More
Bellco Glass, Inc. 24-Well Deep-Well Plates
SDP
Supplier:  Bellco Glass, Inc. 130000317
Encompass_Preferred
24-well Deep-Well Plates have the same footprint as the regular microplates and are available in sterile and non-sterile versionsSuitable for dilution and storageDistinct alpha-numeric well designationsWell volume: 10 mLEach well bottom has rounded corners and chamfered edges to eliminate dead space during mixingPolypropylene lids are availableEZ Seal film compatible48 Deep-well and 96 Deep-well units are also available. Style-Non-sterile, Lid-No Lid, Material-Polypropylene Co-Polymer (PPC), Color-Natural, Package Type-5/Bag, Carton Qty-20
Catalog No. 50-233-0850