Learn More
Bellco Glass, Inc. 24-Well Plates Polystyrene
SDP
Supplier:  Bellco Glass, Inc. 130000311
Encompass_Preferred
24-Well Plates are available non-sterile, sterile, and treated for tissue culture. Sterile and treated plates are gamma-irradiated for sterility and are individually packed with lids.Distinct alpha-numeric well designationsWell volume: 3.5 mLLid sold separately6-well, 12-well, 96-well and 384-well units are also available - Items are not available for shipment to Mexico. Style-Non-Sterile, Lid-No Lid, Material-Polystyrene (PS), Color-Natural, Package Type-10/Bag, Carton Qty-100
Catalog No. 50-233-0488