Learn More
Analytical Sales & Services 24-Well Silicone/PTFE Cap Mat, Prevents Crosstalk, Withstands Vortexing, Units Per Package: 5
Supplier:  Analytical Sales & Services 24777
Encompass
The 24 WELL SILICONE/PTFE CAPMAT 5P is designed to ensure reliable sealing for 24-well collection plates, promoting sample protection and integrity. Its robust construction withstands vigorous vortexing and mechanical disruptions, while preventing crosstalk. This advanced cap mat enhances laboratory workflows by maintaining sample purity and stability during homogenization processes, distinguishing it from standard alternatives.
Catalog No. NC1129062