Learn More
STEMCELL Technologies 245 mm Square Dish, Tissue Culture-Treated, Size: 16 Units
SDP
Supplier:  STEMCELL Technologies 38039
Encompass
245 mm Square Dish Tissue Culture-Treated is ideal for attachment-dependent cell and tissue culture. TC-treated polystyrene is clear and transparent for superior optical clarity. These dishes are sterilized ready for use in an aseptic environment and vented to allow for gas exchange. These dishes are also stackable for easy handling and storage.
Catalog No. 50-239-0156