Learn More
Medchemexpress LLC 24R-Calcipotriol | 112827-99-3 | C27H40O3 | 1 MG
SDP
Supplier:  Medchemexpress LLC HY152661MG
Encompass_Preferred
24R-Calcipotriol (PRI 2202) is an impurity of Calcipotriol, a ligand of VDR-like receptors and a Vitamin D3 analog. It displays minimal effects on calcium homeostasis and regulates cell differentiation and proliferation.
  • Exhibits antiproliferative activity against human HL-60, HL60/MX2, MCF-7, T47D, SCC-25, and mouse WEHI-3 cancer cell lines.
Catalog No. 50-003-72306