Learn More
Analytical Sales & Services 24 x 8mL Aluminum Multi-well Reaction Block (for 2 Dram Vials)
Supplier:  Analytical Sales & Services 24017
Encompass
24-Well Aluminum Reaction Block for 17mm OD (2 Dram) Vials. Includes: Base Plate, Cover, Screws, PFA Film and 2 Rubber Mats.
Catalog No. NC1565243