Learn More
Medchemexpress LLC 25,26-dihydroxyvitamin D3 | 29261-12-9 | 95.5% | 416.64 g/mol | C27H44O3 | 10 MG
SDP
Supplier:  Medchemexpress LLC HY1583010MG
Encompass_Preferred
25,26-Dihydroxyvitamin D3 is a research-grade metabolite of vitamin D3 used to study vitamin D metabolism and intestinal calcium transport. The compound is supplied as a white to off-white solid, with reported purity around 95.5%, and should be stored protected from light and under inert atmosphere for best stability.
  • Metabolite of vitamin D3 with intestinal calcium transport activity.
  • Applicable to studies of vitamin D metabolism and signaling.
  • Reported purity 95.5%.
  • Solid, white to off-white physical form.
  • Storage: protect from light and store under inert atmosphere; in solvent, keep at -80°C for long-term.
  • Intended for research use only.
Catalog No. 50-002-87957
Add to Cart
Request a Quote