Learn More
Chemglass Life Sciences Replacement 25.4mm Stirrer Shaft, Reactor Size: 50L Squatty/400mm Flange, Fits Support Frame: CG-1968-Y-12, Overall Length: 43-3/8"
SDP
Supplier:  Chemglass Life Sciences CG209758
Encompass_Preferred
Replacement 25.4mm Stirrer Shaft, Reactor Size: 50L Squatty/400mm Flange, Fits Support Frame: CG-1968-Y-12, Overall Length: 43-3/8". Precision ground and polished stirrer shaft for use with Chemglass process reactors. Designed to utilize CG-2095 and CG-2096 PTFE agitators. Lower portion of stirrer shaft is sandblasted and has a 3/8 wide flat. REPLACEMENT STIRRER SHAFTS ARE FOR USE WITH MECHANICAL SEAL STIR BEARING ON LISTED SUPPORT FRAME.
Catalog No. 50-153-1451