Learn More
Chemglass Life Sciences Optional 25.4mm Stirrer Shaft, Reactor Size: 50L (Short), Fits Support Frame: CG-1968-F-10 & CG-1968-F-12, Overall Length: 39.6"
SDP
Supplier:  Chemglass Life Sciences CG209750
Encompass_Preferred
25.4mm Stirrer Shaft, Reactor Size: 50L (Short), Fits Support Frame: CG-1968-F-10 & CG-1968-F-12, Overall Length: 39.6". Precision ground and polished stirrer shaft for use with Chemglass process reactors. Designed to utilize CG-2095 and CG-2096 PTFE agitators. Lower portion of stirrer shaft is sandblasted and has a 3/8¿ wide flat. REPLACEMENT STIRRER SHAFTS ARE FOR USE WITH MECHANICAL SEAL STIR BEARING ON LISTED SUPPORT FRAME.
Catalog No. CG209750