Learn More
Chemglass Life Sciences Replacement 25.4mm Stirrer Shaft, Reactor Size: 30L, Fits Support Frame: CG-1968-X-12, Overall Length: 41.9"
SDP
Supplier:  Chemglass Life Sciences CG209731
Encompass_Preferred
Replacement 25.4mm Stirrer Shaft, Reactor Size: 30L, Fits Support Frame: CG-1968-X-12, Overall Length: 41.9". Precision ground and polished stirrer shaft for use with Chemglass process reactors. Designed to utilize CG-2095 and CG-2096 PTFE agitators. Lower portion of stirrer shaft is sandblasted and has a 3/8¿ wide flat. REPLACEMENT STIRRER SHAFTS ARE FOR USE WITH MECHANICAL SEAL STIR BEARING ON LISTED SUPPORT FRAME.
Catalog No. 50-872-527