Learn More
PHCbi VIP ECO SMART Natural Refrigerant 18.6 Cu. Ft. Ultra-Low Temp Freezer (-86°C) | 115V | Capacity: 384 x 2″ Boxes
Supplier:  PHCbi MDFDU503VHAPA
Encompass_Preferred
Independently tested and documented, the MDF-DU503VHA-PA ENERGY STAR Certified PHCbi brand VIP ECO SMART delivers the lowest daily energy usage of any ultra-low temperature freezer of its size: 4.99 kWh per day. The dual voltage ULT freezer includes both a 115V and 220V power cord that can be selected, easily attached when the freezer is installed, or switched to the other voltage upon relocation if desired. Purpose-designed for the temperature uniformity and stability required for the life sciences.
Catalog No. 50-274-0461
Add to Cart
Request a Quote