Learn More
Medchemexpress LLC 25-desacetyl rifampicin | 16783-99-6 | >95.0% | 780.90 | C41H56N4O11 | 1MG
SDP
Supplier:  Medchemexpress LLC HY1255871MG
Encompass_Preferred
25-Desacetyl rifampicin is the primary deacetylated metabolite of rifampicin, supplied as a research-grade analytical reference standard for use in bioanalytical and pharmacokinetic studies. It is intended for research use only and is used as a tracer or internal standard in quantitative methods.
  • Used as an analytical reference standard and metabolite control.
  • Suitable as a tracer or internal standard for NMR, GC-MS, and LC-MS.
  • Molecular formula C41H56N4O11 and molecular weight 780.90.
  • High purity for analytical workflows (>95% reported by some suppliers).
  • Supplied in small milligram quantities appropriate for reference and tracer studies.
Catalog No. 50-002-35707