Learn More
Medchemexpress LLC 25-desacetyl rifampicin | 16783-99-6 | MFCD08063717 | >95.0% | 780.90 | C41H56N4O11 | 5MG
SDP
Supplier:  Medchemexpress LLC HY1255875MG
Encompass_Preferred
25-Desacetyl rifampicin is the deacetylated metabolite of rifampicin provided as an isotope-labeled (deuterated) reference standard for research use. It is intended for analytical quantitation and ADME/pharmacokinetic studies as a tracer or internal standard. The compound has CAS 16783-99-6, molecular formula C41H56N4O11, and molecular weight 780.90 g/mol. Documentation including a data sheet and safety data sheet is available from the supplier.
  • Deuterated isotope-labeled reference standard.
  • Suitable for pharmacokinetic and metabolic studies.
  • High purity (≥95%).
  • Molecular formula C41H56N4O11; molecular weight 780.90 g/mol.
  • Available in small research pack sizes (1 mg-50 mg).
  • Data sheet and safety data sheet available.
Catalog No. 50-002-37322