Learn More
Medchemexpress LLC 25-Hydroxy VD2-d6 | 1262843-46-8 | C28H38D6O2 | 1 MG
SDP
Supplier:  Medchemexpress LLC HY153281MG
Encompass_Preferred
25-Hydroxy VD2-d6 is the deuterium labeled 25-Hydroxy VD2.
  • Can be used as a tracer.
  • Can be used as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS.
Catalog No. 50-003-73607