Learn More
Selleck Chemical LLC 25-Hydroxy vitamin D2-d3 E8246-1MG
SDP
Supplier:  Selleck Chemical LLC E82461MG
Encompass_Preferred
25-Hydroxy vitamin D2-d3 is the deuterated form of (22Z)-25-OH-D2 (22Z)-25-OH-D2 is a metabolic intermediate of vitamin D and is produced by hydroxylation in the liver
Catalog No. 50-000-41529