Learn More
Abcam 25-Hydro x yvitamin D3 (Calcifediol), metabolite of vitamin D3, 5MG
Supplier:  Abcam AB1427665MG
Encompass_Preferred
MW 400.6 Da, Purity >98%. Principal cholecalciferol (ab143594) metabolite. Downregulates TLR4 and JAK1/STAT3 signaling. Hydroxylates to calcitriol (ab141456) *in vivo*. Anti-inflammatory effects. Orally active.
The product is subject to the following: Abcam Restricted Use Statement
Catalog No. 50-270-6187