Learn More
Cytiva 25 mm In-line Filter Holder, Delrin Plastic - 25 mm, in-line (6/pkg)
Supplier:  Cytiva 1109
Encompass
These 25 mm in-line filter holders are ideal for liquid or gas filtration.•Ideal for liquid or gas filtration•Lightweight•For low pressure applicationsThe filter holder in Delrin plastic offers broad chemical compatibility and material strength. Likewise the stainless steel filter holder features durable construction with excellent chemical resistance and can withstand high operating temperatures and pressures.These are lightweight and useful for low pressure applications such as in-line air cleaning, liquid filtering, or particulate sampling. This product is not approved for use in medical, clinical, surgical or other patient applications.
Catalog No. NC1480261