Learn More
MOLECULAR DEPOT LLC 25-OH Vit D3 AP - 0.1 mg
Supplier:  MOLECULAR DEPOT LLC B2010614
Encompass_Preferred
25-OH Vitamin D3 Alkaline Phosphatase Conjugate consists of Alkaline Phosphatase (AP ALP) conjugated to 25-OH Vitamin D3 This product has been used in the study of the metabolism of vitamin D as well as the development of Vitamin D detection methods For in vitro research use only
Catalog No. 50-000-13434