Learn More
Abcam 25(OH) Vitamin D ELISA kit, 1 x 96 Tests
Supplier:  Abcam AB2139661X96TESTS
Encompass_Preferred
The 25(OH) Vitamin D ELISA kit is a competitive ELISA designed to quantify 25-hydroxy vitamin D (vitamin D2 and vitamin D3) in serum and plasma with a sensitivity of 1.98 ng/mL. - Wide dynamic range – quantifies 0.5–1010 ng/mL - Assay platform – pre-coated microplate (12 x 8-well strips) for easy setup and reliable results - Fast assay time – complete results in 1 hour 30 minutes - Cited in over 15 publications
The product is subject to the following: Abcam Restricted Use Statement
Catalog No. NC1784929