Learn More
Genscript Corporation 25-OH Vitamin D3 BSA Conjugat
Supplier:  Genscript Corporation T001010.1
Encompass
Vitamin D plays an important role in calcium homeostasis and metabolism Vitamin D3 and vitamin D2 are two forms of vitamin D in human body both of which only a few foods contain The deficiency of vitamin D in human body can cause a threat of diseases such as osteomalacia rickets and diabetes 25-OH Vitamin D2 and 25-OH Vitamin D3 as vitamin D metabolites are measured in human serum to evaluate vitamin D level Vitamin D supplements are considered as an effective way to treat or to prevent the diseases
Catalog No. 50-000-56737