Learn More
REVMAB BIOSCIENCES USA INC 25-OH Vitamin D3 [RM3], Rabbit Monoclonal Antibody
Supplier:  REVMAB BIOSCIENCES USA INC 31117000
Encompass_Preferred
Anti-25-OH Vitamin D3 Rabbit Monoclonal Antibody, Clone RM3(100uL) , not cross-react with Vitamin D3, Vitamin D2, and 25-OH Vitamin D2; Product info: www.revmab.com
Catalog No. NC1728994