Learn More
Innovative Reseach Of America NHH-115 17β-Estradiol Pellets, Consistent Hormone Levels, 60-Day Release, 25 Pellets
SDP
Supplier:  Innovative Reseach Of America NHH115
Encompass
The NHH-115 by Innovative Research of America is a 17β-Estradiol pellet system for controlled hormone delivery in scientific research. Each pellet delivers a precise dose of 0.72 mg over 60 days, using a Matrix-Driven Delivery (MDD) system for subcutaneous implantations. This design ensures stable hormone levels, eliminates the need for intermittent injections, and enhances reproducibility in research, making it ideal for endocrinology, oncology, and reproductive biology studies.
Catalog No. NC3376648