Learn More
Chemglass Life Sciences 250mL 3-Neck European Flask, 1-CN 24/40 Outer, 2-SN 24/40 Outer Angled 20°
SDP
Supplier:  Chemglass Life Sciences CG157006
Encompass_Preferred
250mL 3-Neck European Flask, 1-CN 24/40 Outer, 2-SN 24/40 Outer Angled 20¿. European style flask with tapered walls and a shallow hemispherical bottom for more efficient mechanical or magnetic stirring. With standard taper outer joints on center and side necks.
Catalog No. CG157006