Learn More
Ace Glass, Inc. Beaker, 250mL, jacketed, Ace-Safe hose connections for 1/4intubing, 89mm inside height, 65mm inside diameter
SDP
Supplier:  Ace Glass, Inc. 5340105
Encompass_Preferred
250mL jacketed beaker with one upper and one lower Ace-Safe hose connection on opposite sides, for 1/4 in. tubing. Dimensions: 89mm I.H. x 65mm I.D. Does not have a pouring spout.
Catalog No. 50-971-848