Learn More
Chemglass Life Sciences Cylinder, Economy, 250mL, Graduated, Double Scale
SDP
Supplier:  Chemglass Life Sciences CG8246250
Encompass_Preferred
Cylinder, Economy, 250mL, Graduated, Double Scale Calibrated To Contain. A less costly cylinder, designed for those institutions performing many general laboratory procedures. No bumper guards are supplied.
Catalog No. NC0192096