Learn More
Ace Glass, Inc. 250mL gas washing bottle with porosity C (25-50 micron) fritted disc and 29/42 joint. Inlet/outlet arms are 8mm O.D.
SDP
Supplier:  Ace Glass, Inc. 716664
Encompass_Preferred
250mL gas washing bottle with porosity C (25-50 micron) fritted disc and 29/42 joint. Inlet/outlet arms are 8mm O.D.
Catalog No. 50-974-140