Learn More
LAB261 250µm Plain Blue Color Dyed Polystyrene Microspheres, 5ml
Supplier:  LAB261 DBP250K
Encompass
Color Dyed Polystyrene microspheres are suitable to enhance both visual and microscopic identification. Our color dyed microspheres are frequently used in lateral flow tests and latex agglutination tests. They are available with non-functionalized surfaces and with carboxyl functionalized surfaces to support covalent coupling strategies. These surface modified microspheres are suitable for the covalent attachment of proteins, peptides, and nucleic acids.
Catalog No. NC3781507