Learn More
Chemglass Life Sciences Replacement 254 nm UV Bulb for Chemglass CLS-1625 Handheld UV Lamp
SDP
Supplier:  Chemglass Life Sciences CLS1625254
Encompass
Chemglass replacement bulb that emits 254 nm ultraviolet (UV-C) light, designed as a direct replacement for Chemglass handheld UV lamps in the CLS-1625 product family. Sold individually, it provides a 254 nm output for UV-based procedures.
  • Emits 254 nm ultraviolet (UV-C) light.
  • Designed as a direct replacement for handheld UV lamps.
  • Sold individually.
  • Provides a consistent 254 nm output for UV-based procedures.
Catalog No. NC3235041