Learn More
Cayman Chemical 25C-NBFhydrochlorIde 10mg
SDP
Supplier:  Cayman Chemical 1596910 MG
Encompass_Preferred
A derivative of 2C-C having an N-(2-fluorobenzyl) addition at the amine; an agonist of serotonin receptors that activates 5-HT2A and 5-HT2C similarly (EC50s = ~0.3 µM for EAch); intended for forensic and research applications
Catalog No. 50-296-8685