Learn More
Bachem Fmoc-Cl
SDP
Supplier:  Bachem 4004776.0025
Encompass_Preferred
Fmoc-Cl, 28920-43-6, C15H11ClO2, Mr 258.70, 25g. Fmoc-chloride. Reagent for the introduction of the Fmoc-amino-protecting group, which is stable towards acids and catalytic hydrogenation but is readily cleaved under mildly basic non-hydrolytic conditions. The use of this reagent in the protection of hydroxy groups and amino groups in nucleotides has been reported as well. Fmoc-chloride is often used for pre-column derivatization of amines and amino acids in analytical HPLC.
Catalog No. NC9468377