Learn More
Cayman Chemical 25hydroxy VItamIn D3 RabbIt
SDP
Supplier:  Cayman Chemical 32179100 UL
Encompass_Preferred
An allosteric inhibitor of FGFR2 inhibits FGF2-induced phosphorylation of FRS2a in KATO III cells (IC50 10 nM) inhibits the growth of SKOV3 and Hs 578T cells (GI50s 370 and 210 nM respectively) inhibits bFGF-induced HUVEC proliferation (IC50 11 nM) inhibits SVEC4-10 cell tube formation at 50 nM decreases the number of tumor microvessels by 50 in a SKOV3 ovarian serous carcinoma mouse model reduces tumor growth in mouse xenograft models at 30 mg/kg per day
Catalog No. 50-294-9370